Learn More
Chemglass Life Sciences 3000mL Erlenmeyer Flask, 38-430 GPI Screw Thread
SDP
Catalog No. 50873164
Encompass_Preferred
3000mL Erlenmeyer Flask, 38-430 GPI Screw Thread. Erlenmeyer flask with screw thread finish. Supplied complete with white rubber lined black phenolic cap. Long skirted caps can be loosened without falling off while shaking flask.
Catalog No. 50-873-164 Supplier Chemglass Life Sciences Supplier No. CG154307
3000mL Erlenmeyer Flask, 38-430 GPI Screw Thread. Erlenmeyer flask with screw thread finish. Supplied complete with white rubber lined black phenolic cap. Long skirted caps can be loosened without falling off while shaking flask.