Learn More
Chemglass Life Sciences 3, 000mL Glassblowers Heavy Wall RBF Blank
SDP
Catalog No. 501531801
Encompass_Preferred
3,000mL Glassblowers Heavy Wall Round Bottomed Flask Blank. Heavy Wall blanks are 30% heavier than Chemglass's standard CG-618 series. Blanks are used in the fabrication of single or multi-neck round bottom flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.
Catalog No. 50-153-1801 Supplier Chemglass Life Sciences Supplier No. CG61812HW
3,000mL Glassblowers Heavy Wall Round Bottomed Flask Blank. Heavy Wall blanks are 30% heavier than Chemglass's standard CG-618 series. Blanks are used in the fabrication of single or multi-neck round bottom flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.