Learn More
Bellco Glass, Inc. 3L Heating Blanket,115V Bioreactor
SDP
Catalog No. 501946498
Encompass_Preferred
The blanket has velcro closures for easy setup and a 6" power cable with IEC 60320 C14 plug. 3L Heating Blanket,115V Bioreactor. MADE IN THE USA
Catalog No. 50-194-6498 Supplier Bellco Glass, Inc. Supplier No. 790923115
The blanket has velcro closures for easy setup and a 6" power cable with IEC 60320 C14 plug. 3L Heating Blanket,115V Bioreactor. MADE IN THE USA