Learn More
Alkali Scientific 3L High Efficiency Erlenmeyer
SDP
Catalog No. 503028805
Encompass_Preferred
Our 3L high efficiency shaker flasks are ideal for medium size scale up work They are inhigh efficiencyin in that a 3L flask has 162mm diameter footprint allowing for more of them to fit on a shaker platform The geometry of the flasks also allows for higher shaker speeds without damaging the cells Faster shaker speeds means better gas exchange higher growth rates and more yield per flask This product features pre-assembled tubing connectors and gas exchange filters allowing for the movement of liquid in and out of the Erlenmeyer flask with minimal risk of contamination Many customers have realized significant cost savings by switching to aseptic transfer systems 3L High Efficiency Erlenmeyer Flask 1 system per double wrapped sterile bag - 4 systems per case Flask made of USP grade VI virgin polycarbonate Untreated super smooth interior surface ideal for suspended cultures Attached bi-directional transfer cap Attached thermoplastic elastomer (TPE) external transfer tube Male C
Catalog No. 50-302-8805 Supplier Alkali Scientific Supplier No. EFC622
Our 3L high efficiency shaker flasks are ideal for medium size scale up work They are inhigh efficiencyin in that a 3L flask has 162mm diameter footprint allowing for more of them to fit on a shaker platform The geometry of the flasks also allows for higher shaker speeds without damaging the cells Faster shaker speeds means better gas exchange higher growth rates and more yield per flask This product features pre-assembled tubing connectors and gas exchange filters allowing for the movement of liquid in and out of the Erlenmeyer flask with minimal risk of contamination Many customers have realized significant cost savings by switching to aseptic transfer systems 3L High Efficiency Erlenmeyer Flask 1 system per double wrapped sterile bag - 4 systems per case Flask made of USP grade VI virgin polycarbonate Untreated super smooth interior surface ideal for suspended cultures Attached bi-directional transfer cap Attached thermoplastic elastomer (TPE) external transfer tube Male C