Learn More
Bellco Glass, Inc. 3L Insct Sys Digital IOB,115V
SDP
Catalog No. 501945076
Encompass_Preferred
Designed for growing insect cell cultures such as SF9 and SF21 cells. The system comes completewith all accessories needed to grow your insect culture line for expression. MADE IN THE USA Includes:–7785-D2005 Digital 5-Position Magnetic Stirrer 115V–1965-03000 Micro-Carrier Spinner Flask Complete 3L–1965-20003R 2-Port Assembly 3L–7808-10004 Gas Blending Stand–7725-00120 Air Supply Pump 115V–7800-10050 Silicone Tubing 1/4” ID
Catalog No. 50-194-5076 Supplier Bellco Glass, Inc. Supplier No. 196576115
Add to Cart
Add to Cart
Request a Quote
Designed for growing insect cell cultures such as SF9 and SF21 cells. The system comes completewith all accessories needed to grow your insect culture line for expression. MADE IN THE USA Includes:–7785-D2005 Digital 5-Position Magnetic Stirrer 115V–1965-03000 Micro-Carrier Spinner Flask Complete 3L–1965-20003R 2-Port Assembly 3L–7808-10004 Gas Blending Stand–7725-00120 Air Supply Pump 115V–7800-10050 Silicone Tubing 1/4” ID