Learn More
Bellco Glass, Inc. 3L Insct Sys Digital MicroCarrier ,230V
SDP
Catalog No. 501945074
Encompass_Preferred
Designed for growing insect cell cultures such as SF9 and SF21 cells. The system comes completewith all accessories needed to grow your insect culture line for expression. MADE IN THE USA Includes:–Digital 5-Position Magnetic Stirrer 230V–1965-03000 Micro-Carrier Spinner Flask Complete 3L–1965-20003R 2-Port Assembly 3L–7808-10004 Gas Blending Stand–Air Supply Pump 230V–7800-10050 Silicone Tubing 1/4” ID
Catalog No. 50-194-5074 Supplier Bellco Glass, Inc. Supplier No. 196570230
Add to Cart
Add to Cart
Request a Quote
Designed for growing insect cell cultures such as SF9 and SF21 cells. The system comes completewith all accessories needed to grow your insect culture line for expression. MADE IN THE USA Includes:–Digital 5-Position Magnetic Stirrer 230V–1965-03000 Micro-Carrier Spinner Flask Complete 3L–1965-20003R 2-Port Assembly 3L–7808-10004 Gas Blending Stand–Air Supply Pump 230V–7800-10050 Silicone Tubing 1/4” ID