Learn More
Bellco Glass, Inc. 3L Insect Sys Digital MicroCarrier ,115V
SDP
Catalog No. 501945073
Encompass_Preferred
Designed for growing insect cell cultures such as SF9 and SF21 cells. The system comes complete with all accessories needed to grow your insect culture line for expression. MADE IN THE USA.Consist Of:7785-D2005 Digital 5-Position Magnetic Stirrer, 115V1965-03000 Micro-Carrier Spinner Flask Complete, 3L1965-20003R 2-Port Assembly, 3L7808-10004 Gas Blending Stand7725-00120 Air Supply Pump, 115V7800-10050 Silicone Tubing, 1/4″ ID
Catalog No. 50-194-5073 Supplier Bellco Glass, Inc. Supplier No. 196570115
Add to Cart
Add to Cart
Request a Quote
Designed for growing insect cell cultures such as SF9 and SF21 cells. The system comes complete with all accessories needed to grow your insect culture line for expression. MADE IN THE USA.Consist Of:7785-D2005 Digital 5-Position Magnetic Stirrer, 115V1965-03000 Micro-Carrier Spinner Flask Complete, 3L1965-20003R 2-Port Assembly, 3L7808-10004 Gas Blending Stand7725-00120 Air Supply Pump, 115V7800-10050 Silicone Tubing, 1/4″ ID