Learn More
Chemglass Life Sciences 3L Media Transfer Vessel, Baffled Bottom, Barbed Fitting, Weldable C-Flex Tubing, Plugged, Sterile, Complete
SDP
Catalog No. 5000083164
Encompass
Media Transfer Vessel is ideal for use when doing a gravity feed from a sterile 3L vessel.Fernbach flasks are sterile and individually double bagged.The 3L polycarbonate flasks with filter caps have a 3/16" barbed fittings that accept an 1/8" ID, 1/4" OD weldable C-Flex tubing end plugged.Supplied complete with a Tubing Clamp.Gamma irradiated Sterile.Please call for bulk pricing.
Catalog No. 50-000-83164 Supplier Chemglass Life Sciences Supplier No. CLS11713LB
Media Transfer Vessel is ideal for use when doing a gravity feed from a sterile 3L vessel.Fernbach flasks are sterile and individually double bagged.The 3L polycarbonate flasks with filter caps have a 3/16" barbed fittings that accept an 1/8" ID, 1/4" OD weldable C-Flex tubing end plugged.Supplied complete with a Tubing Clamp.Gamma irradiated Sterile.Please call for bulk pricing.