Learn More
Chemglass Life Sciences Flask, Round Bottom, 3000mL, Heavy Wall, 3-Neck, 29/42 CN - 24/40 SN's Angled 20°
SDP
Catalog No. CG152415
Encompass_Preferred
Flask, Round Bottom, 3000mL, Heavy Wall, 29/42 - 24/40, 3-Neck, Angled 20¿. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20¿ angled side necks.Center neck: 29/42Side neck: 24/40
Catalog No. CG152415 Supplier Chemglass Life Sciences Supplier No. CG152415
Flask, Round Bottom, 3000mL, Heavy Wall, 29/42 - 24/40, 3-Neck, Angled 20¿. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20¿ angled side necks.Center neck: 29/42Side neck: 24/40