Learn More
Chemglass Life Sciences Flask, Round Bottom, 3000mL, Heavy Wall, Vertical 4-Neck, 34/45 CN, 24/40 SN's
SDP
Catalog No. CG153041
Encompass_Preferred
Flask, Round Bottom, 3000mL, Heavy Wall, 34/45 - 24/40, 4-Neck, Vertical. Round bottom, HEAVY WALL, four neck flask with standard taper outer joints on the center and vertical side necks.
Catalog No. CG153041 Supplier Chemglass Life Sciences Supplier No. CG153041
Flask, Round Bottom, 3000mL, Heavy Wall, 34/45 - 24/40, 4-Neck, Vertical. Round bottom, HEAVY WALL, four neck flask with standard taper outer joints on the center and vertical side necks.