Learn More
Ace Glass, Inc. Flask, round bottom, heavy wall, four neck, 3L, 45/50 center, 24/40 sides
SDP
Catalog No. NC9462453
Encompass_Preferred
3L round bottom, heavy wall flask with four necks, arranged three in-line and one at 90. Center neck has a 45/50 joint, and angled side necks have 24/40 joints.
Catalog No. NC9462453 Supplier Ace Glass, Inc. Supplier No. 6952244
3L round bottom, heavy wall flask with four necks, arranged three in-line and one at 90. Center neck has a 45/50 joint, and angled side necks have 24/40 joints.