Learn More
Alkali Scientific 3L Wide-mouth High Efficiency
SDP
Catalog No. 503027421
Encompass_Preferred
The high-efficient Erlenmeyer flask (with tube) is designed to eliminate pipeline design assembly sterilization and other operations and improving production efficiency It is ideal for shaker culture applications and can be used for scale-up culture of suspension cells Polycarbonate HDPE cap Bi-directional transfer cap with TPE tube Handle Nylon Sterilized by E-beam Sterility assurance level SAL 10 Non-pyrogenic and DNase/RNase free Pipe diameter / in Int / in Ext High-efficiency large-volume culture flasks allow cells to show strong viability with large expression amount of proteins in the culture of mammalian cells and insect cells During the culturing process the use rate of the shaker is significantly increased and the survival rate and viability of cells are both dramatically It is pre-installed on the High efficiency Erlenmeyer flask which realizes the closed transfer of liquid and reduces the risk of external contamination during the process of capping and screwing T
Catalog No. 50-302-7421 Supplier Alkali Scientific Supplier No. EFC624
The high-efficient Erlenmeyer flask (with tube) is designed to eliminate pipeline design assembly sterilization and other operations and improving production efficiency It is ideal for shaker culture applications and can be used for scale-up culture of suspension cells Polycarbonate HDPE cap Bi-directional transfer cap with TPE tube Handle Nylon Sterilized by E-beam Sterility assurance level SAL 10 Non-pyrogenic and DNase/RNase free Pipe diameter / in Int / in Ext High-efficiency large-volume culture flasks allow cells to show strong viability with large expression amount of proteins in the culture of mammalian cells and insect cells During the culturing process the use rate of the shaker is significantly increased and the survival rate and viability of cells are both dramatically It is pre-installed on the High efficiency Erlenmeyer flask which realizes the closed transfer of liquid and reduces the risk of external contamination during the process of capping and screwing T