Learn More
Electron Microscopy Sciences Lyophilization Three Leg Stoppers Gray, 13 mm I.D., 20 mm O.D.
SDP
Catalog No. 501898955
Encompass_Preferred
Manufactured from the highest quality raw materials. Fits mouth with:
I.D.: 13 mm
O.D.: 20 mm
Material: Gray holobutyl/50
Catalog No. 50-189-8955 Supplier Electron Microscopy Sciences Supplier No. 6032713
Manufactured from the highest quality raw materials. Fits mouth with:
I.D.: 13 mm
O.D.: 20 mm
Material: Gray holobutyl/50