Learn More
Electron Microscopy Sciences Lyophilization Three Leg Stoppers for 13mm ID, 20 mm O.D.
SDP
Catalog No. 501898956
Encompass_Preferred
Manufactured from the highest quality raw materials. Fits mouth with:
I.D.: 13 mm
O.D.: 20 mm
Material: Gray chlorobutyl/50
Catalog No. 50-189-8956 Supplier Electron Microscopy Sciences Supplier No. 6032714
Manufactured from the highest quality raw materials. Fits mouth with:
I.D.: 13 mm
O.D.: 20 mm
Material: Gray chlorobutyl/50