Learn More
3M™ 1600-Series Safety Sunwears
Catalog No. 17623759 Shop All 3M Products
Change view
Click to view available options
Anti-scratch coated lens; full-frame; Black frame; Red mirror lens
Catalog No.
17-623-759 Anti-scratch coated lens; full-frame; Black frame; Red mirror lens
1 options
This item is not returnable. View return policy
Catalog No. 17-623-759 Supplier 3M™ Supplier No. 7010341171
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
The sleek 1600-Series unites comfort, coverage and clarity. Wraparound frame features soft nose pads, impact-resistant polycarbonate lenses and color-accented temples.
3M™ Safety Sunwear Protective Eyewear helps protect eyes from dust, debris and the harmful rays of the sun, yet look bold and contemporary for a stylish appearance. These safety eye glasses combine the protection you need with the appearance you want.
  • On-the-job eye protection with a sleek and bold design
  • Removable foam liners offer extra comfort and protection from nuisance dust
  • Polycarbonate lenses absorb 99.9% UVA and UVB up to 380 nm
  • Meets the requirements of CSA Z94.3 and ANSI Z87.1-2010
  • 3M Catalog# SS1629AS-B
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Red
Product Type Safety Glasses
Frame Color Black
Model SS1629AS-B
Temple Color Black
Lens Material Polycarbonate
Frame Material Plastic
UV Protection 0.999
Frame Type Full Frame