Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Hooded Protective Coveralls 4510 Series, Elastic Wrist and Ankles

Catalog No. 17623335 Shop All 3M Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-623-335 Medium
17-623-336 Large
17-623-337 X-Large
17-623-338 2X-Large
17-623-339 3X-Large
17-623-340 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-623-335 Supplier 3M™ Supplier No. 7000089662
Add to Cart
Add to Cart
This item is not returnable. View return policy

Coveralls provide light duty protection against infective agents, certain hazardous dusts, and limited liquid splashes. PE laminate material includes micropores that allow heat and moisture to evaporate from the skin and out through the suit for a lightweight and comfortable fit.

  • Type 5/6 Coverall designed to offer cost effective protection against hazardous dusts and limited liquid splashes
  • Protection against EN 14126 Biological agents
  • Made of a soft lightweight non-woven with microporous laminate for splash protection, strength and breathability
  • Certified protection against Type 5/6 hazardous dusts and limited liquid splashes
  • 2 panel hood design, 2-way zip with storm flap, elasticated apertures reduce the risk of skin exposure
  • Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size Medium
Material Microporous Polyethylene Laminate
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Closure Type 2-way Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Coating Anti-static
Certifications/Compliance EN 13034 Type 6, EN ISO 13982-1 Type 5, EN 1149-1:2006, ASTM D2859-96, CE
Latex Free Yes
Model 4510-M
Size (English) Chest 36 to 39 in.
Size (English) Inseam 30 in.
Size (English) Waist 50 in.
Size (Metric) Chest 91.44 to 99.06 cm
Size (Metric) Inseam 76.2 cm
Size (Metric) Waist 127 cm
For Use With (Application) Abatement, Remediation, Agriculture, Cleaning and Maintenance, General Industry, Manufacturing, Non-Hazardous Operations, Painting, Pharmaceutical Cleanroom
Height (English) 5.5 to 5.75 ft.
Height (Metric) 167 to 176 cm
Cleanroom Class Class 10 (ISO 4)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.