Learn More
Uline 3M 471 Vinyl Tape, Blue
Catalog No. NC1465995
Encompass
Do-it-all tape color codes masks seals splices and marks hazardous or restricted areas. Conforms to curved and irregular surfaces. Maintains color even with heavy abrasion. Handles temperatures from 40F to 170F. Aggressive rubber adhesive Residue-free removal.
Catalog No. NC1465995 Supplier Uline Supplier No. S10253BLU
Do-it-all tape color codes masks seals splices and marks hazardous or restricted areas. Conforms to curved and irregular surfaces. Maintains color even with heavy abrasion. Handles temperatures from 40F to 170F. Aggressive rubber adhesive Residue-free removal.