Learn More
Uline 3M 471 Vinyl Tape - 12 x 36 yds, Blue
Supplier:  Uline S10251BLUPK
Encompass_Preferred
Do-it-all tape color codes masks seals splices and marks hazardous or restricted areas, Conforms to curved and irregular surfaces, Maintains color even with heavy abrasion, Handles temperatures from 40F to 170F, Aggressive rubber adhesive Residue-free removal, 14 LB TENSILE STRENGTH, OSHA Safety Color Compliant
Catalog No. NC1383388