Learn More
Uline 3M 471 Vinyl Tape, Red
Catalog No. NC2108968
Encompass_Preferred
Do-it-all tape color codes masks seals splices and marks hazardous or restricted areas. Conforms to curved and irregular surfaces. Maintains color even with heavy abrasion. Handles temperatures from 40F to 170F. Aggressive rubber adhesive Residue-free removal. 14 LB TENSILE STRENGTH. OSHA Safety Color Compliant.
This item is not returnable. View return policy
Catalog No. NC2108968 Supplier Uline Supplier No. S10252R
Do-it-all tape color codes masks seals splices and marks hazardous or restricted areas. Conforms to curved and irregular surfaces. Maintains color even with heavy abrasion. Handles temperatures from 40F to 170F. Aggressive rubber adhesive Residue-free removal. 14 LB TENSILE STRENGTH. OSHA Safety Color Compliant.