Learn More
Uline 3M 471 Vinyl Tape, White
Catalog No. NC2124901
Encompass
Do-it-all tape color codes masks seals splices and marks hazardous or restricted areas. Conforms to curved and irregular surfaces. Maintains color even with heavy abrasion. Handles temperatures from 40F to 170F. Aggressive rubber adhesive Residue-free removal. OSHA Safety Color Compliant.
This item is not returnable. View return policy
Catalog No. NC2124901 Supplier Uline Supplier No. S10253W
Do-it-all tape color codes masks seals splices and marks hazardous or restricted areas. Conforms to curved and irregular surfaces. Maintains color even with heavy abrasion. Handles temperatures from 40F to 170F. Aggressive rubber adhesive Residue-free removal. OSHA Safety Color Compliant.