Learn More
Uline 3M 4910 VHB Double-Sided Tape, Clear
Supplier:  Uline S10112
Encompass
40 MIL, Replaces rivets bolts screws welding and liquid adhesives, Incredible VHB strength guarantees a strong bond, Bonds transparent materials, Temperature range is -40F to 200F, Dampens noise shock and vibration, Resists moisture UV and solvents, Indoor/outdoor, Clear acrylic with acrylic adhesive, red polyethylene film liner, UL Listing 746C
Catalog No. NC0775180