Learn More
Uline 3M 6""X72YDS TAPE 4PK
Catalog No. NC1696333
Encompass
3m 821 label protectio tape 2.5mil, application above 65f(room temp), performance below freezing to 200f, can be written with pencil, pen, or markers
Catalog No. NC1696333 Supplier Uline Supplier No. S7165PK
3m 821 label protectio tape 2.5mil, application above 65f(room temp), performance below freezing to 200f, can be written with pencil, pen, or markers