Learn More
MSC 3M 6000 SERIES CARTRIDGES
Catalog No. 5000793531
Encompass
3M Bayonet Gas & Vapor Cartridges with Particulate Filters are NIOSH approved and specifically designed to protect against ammonia and methylamine vapors, as well as P100 particulates. Featuring a bayonet-style connection, these cartridges seamlessly integrate with compatible 3M reusable respirators, offering lightweight respiratory protection for hazardous environments while optimizing the user's field of vision.
  • NIOSH approved for protection against ammonia, methylamine, and P100 particulates
  • Bayonet-style connection for compatible reusable respirators
  • Swept-back, low profile design enhances field of view and balance
  • Lightweight with low breathing resistance
  • Quarter turn for quick and easy installation
  • Particulate filter permanently attached to cartridge
Catalog No. 50-007-93531 Supplier MSC Supplier No. 71855241
3M Bayonet Gas & Vapor Cartridges with Particulate Filters are NIOSH approved and specifically designed to protect against ammonia and methylamine vapors, as well as P100 particulates. Featuring a bayonet-style connection, these cartridges seamlessly integrate with compatible 3M reusable respirators, offering lightweight respiratory protection for hazardous environments while optimizing the user's field of vision.
  • NIOSH approved for protection against ammonia, methylamine, and P100 particulates
  • Bayonet-style connection for compatible reusable respirators
  • Swept-back, low profile design enhances field of view and balance
  • Lightweight with low breathing resistance
  • Quarter turn for quick and easy installation
  • Particulate filter permanently attached to cartridge