Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ 6000 Series Drop-Down Half Facepiece

Catalog No. 19041417 Shop All 3M Products
Change view
Click to view available options
For Use With (Application):
Workers who wear hardhats and need to frequently remove the respirator without removing the hardhat
Format:
Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Format
19-041-417 Workers who wear hardhats and need to frequently remove the respirator without removing the hardhat Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-041-417 Supplier 3M™ Supplier No. 6100DD

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Requires

Facepiece requires compatible cartridges and/or filters. (See separate listings.)

Designed for workers who need to communicate frequently

  • Head cradle adjusts to the individual worker's head size
  • Head harness system allows facepiece to drop to the chest without user having to remove hardhat
  • Durable straps hold their elasticity for long wear
  • Upper strap adjustment is on the head cradle so the strap ends do not interfere with worker's vision
  • Exhalation valve cover directs air downward to help prevent fogging of the faceshield

These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.

Order Info

Can be used with Series 2000 or 7093 filters or Series 6000 cartridges. Contact your Fisher Safety Sales Representative for compatibility information related to various applications.

Certifications

NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NIOSH 42CFR84
Connection Type Bayonet
Material Thermoplastic Elastomer
Product Type Drop Down Half Facepiece Respirator
Format Small
For Use With (Application) Workers who wear hardhats and need to frequently remove the respirator without removing the hardhat
For Use With (Equipment) Filter 2000 series, filter 7093, p100 or cartridge 6000 series
Model 6100DD
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.