Learn More
3M™ 7500 Series Reusable Half Facepiece Reusable Respirators
Half-mask respirators with enhanced comfort and visibility use a bayonet attachment system for twin lightweight filters to protect against gases, vapors and particulates.
Supplier:  3M™ 7100134948
 View more versions of this product
Catalog No. 19-096-898
Edge
Add to Cart
Alert:
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Requires : Compatible cartridges and/or filters. Accommodates 3M 7093 and Series 2000 filters and Series 6000 cartridges.
Description
Description
  • Durable, reusable and heat-resistant mask is made of advanced silicone material.
  • Unique adjustment design helps reduce tension and pressure points on the face.
  • Cool Flow™ exhalation valve opens easier, lowering exhalation breathing resistance, reducing heat and moisture buildup.
  • Easily adjusted, dual-mode head harness gives option of wearing in traditional or drop-down mode.
  • Ideal choice for welding applications.
  • Color-coded sizes for easy identification.
  • 3M Catalog# 7501, 3M Catalog# 7502, 3M Catalog# 7503
Specifications
Specifications
NIOSH 42 CFR Part 84
Blue
Half-Mask Respirator
Silicone
Cartridge 6000 series, filter 2000, 2200, 7000 or 5000 series, or dual airline system
4-point
Bayonet
Silicone
Small
Assembly, cleaning, grinding, machine operation, painting, remediation, welding
7501
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Accessories