Learn More
3M™ 8000 N95 Particle Respirator
Catalog No. 19065671 Shop All 3M Products
Change view
Click to view available options
Particulate respirator; Adjustable noseclip
Catalog No.
19-065-671 Particulate respirator; Adjustable noseclip
1 options
Do not exceed maximum use concentration established by regulatory standards. If respirator becomes damaged, soiled, or breathing becomes difficult, leave the contaminated area and dispose of the respirator. Before each use of the product, please inspect for any rips, damage, tears, etc. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Catalog No. 19-065-671 Supplier 3M™ Supplier No. 8000
Do not exceed maximum use concentration established by regulatory standards. If respirator becomes damaged, soiled, or breathing becomes difficult, leave the contaminated area and dispose of the respirator. Before each use of the product, please inspect for any rips, damage, tears, etc. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Patented filter media with advanced electrostatically charged microfibers helps make breathing easier and cooler for enhanced user comfort
3M Filtering Facepiece Particulate Respirators (FFRs) meet NIOSH 42 CFR 84 standards. Recommended for use up to 10X the OSHA PEL or applicable occupational exposure limits, whichever is lower.
Application
  • An economical solution for light-duty or short-duration applications (grinding , sanding, sweeping, other dusty operations)
  • Design
    • Two-strap design with single-point attachment helps provide a secure fit
    • Standard adjustable nose clip reduces eyewear fogging and helps ensure better seal
    Compliance
    N95 approved respirator meets NIOSH 42CFR84 requirements. NIOSH Approval No.: TC-84A-3981.
    TRUSTED_SUSTAINABILITY
    Specifications
    Aerosol Type N - Not Resistant to Oil
    Certifications (NIOSH) NIOSH 42 CFR 84, N95
    Design Type Cup/Molded
    Filter Type N95
    Product Type General Purpose Particulate Respirator
    With Exhalation Valve No
    Model 8000
    FDA Cleared Surgical Mask No