Learn More
3M™ Accessories for Dual Airline Supplied Air Respirators
Catalog No. 19813789 Shop All 3M Products
Change view
Click to view available options
Product Type:
Air Regulating Valve Assembly
Airline Connector Valve Assembly
Includes:
Air Regulator, Waist Belt
Dual Airline Connector, Waist Belt
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Includes
19-813-789 Air Regulating Valve Assembly Air Regulator, Waist Belt
19-813-790 Airline Connector Valve Assembly Dual Airline Connector, Waist Belt
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-813-789 Supplier 3M™ Supplier No. 7000002002
This item is not returnable. View return policy
Includes
waist belt
For use with dual airline supplied air respirators.
  • Automotive, food and beverage manufacturing
  • Metal production and fabrication
  • Military maintenance, repair and operation
  • Pharmaceutical
  • Transportation
  • 3M Catalog# SA-1007 , 3M Catalog# SA-1027
TRUSTED_SUSTAINABILITY
Specifications
Applications Assembly, Cleaning, Electrical, Facility Maintenance, Grinding, Machine Operation, Painting, Sanding, Welding
For Use With (Equipment) Dual Airline Supplied Air Respirator
Includes Air Regulator, Waist Belt
Model SA-1007
Quantity 1 Ea.
Product Type Air Regulating Valve Assembly