Learn More
3M™ Headgear Accessory, Forehead Comfort/Sweat Pad
Catalog No. 19038920 Shop All 3M Products
Change view
Click to view available options
Versaflo M-Series headgear; Forehead comfort/Sweat pad
Catalog No.
19-038-920 Versaflo M-Series headgear; Forehead comfort/Sweat pad
1 options
Catalog No. 19-038-920 Supplier 3M™ Supplier No. 7100043088
  • $102.80
    $80.65 / Case of 10
    Save $22.15
    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Forehead Comfort/Sweat Pad for Headgears
  • For use with Headgears
  • 3M Catalog# M-957/L-115
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
Product Type Comfort Pad
For Use With (Application) Attaches to The Headband in The Area of The Users Forehead, Used in Assembly, Cleaning, Electrical, Facility Maintenance
For Use With (Equipment) Versaflo™ M-Series Headgear, L-Series Headgear, H-Series Hood, and Whitecap™ Helmets.
Model M-957/L-115
Quantity 10/Cs.