Learn More
3M™ Accessories and Replacement Parts for 6000 Series Full Facepieces, Tinted Lens Cover
Catalog No. 19813726 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Full facepiece 6000 series
Catalog No. For Use With (Equipment)
19-813-726 Full facepiece 6000 series
1 options
Catalog No. 19-813-726 Supplier 3M™ Supplier No. 7000052088
Add to Cart
Add to Cart
For use with 6000 Series full facepieces. Not for use in welding or other applications that require reduction of UV or IR radiation beyond the capability of normal nonprescription sunglasses.
  • Accessory to shield eyes from bright, intense light (not for welding)
  • Tinting relaxes eyes to make it easier to focus on outdoor work
  • Designed for use with 3M Full Facepiece 6000 Series
  • Cover fits easily over lens
  • Approved 3M protection accessory
  • 3M Catalog# 6886
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tinted Lens Cover
For Use With (Equipment) Full facepiece 6000 series
Model 6886