Learn More
Enterprise Technology Solutions 3M Adhesive Spray, Photo Mount, 10.3 oz, Clear
SDP
Supplier:  Enterprise Technology Solutions MMM6094
Encompass_Preferred
Scotch Photo Mount Spray Adhesive is a strong, clear professional-grade adhesive developed especially for mounting photos quickly, easily and permanently. It is a clear, pH neutral, nonyellowing adhesive that allows 3 minutes of open time before bonding so you have time to position and reposition prints. Spray adhesive is also excellent for mounting art prints, maps, illustrations and more.
Catalog No. 50-008-29009