Learn More
3M™ 6000 / 6000DIN / 7000 Series Full Facepiece Accessory, Air Regulating Kit
Catalog No. 19021566 Shop All 3M Products
Catalog No. 19-021566 Supplier 3M™ Supplier No. 7010393905
Air regulating kit for use with full facepieces.
  • 3M Catalog# W-3197
TRUSTED_SUSTAINABILITY
Specifications
Applications Assembly, Blasting, Grinding, Machine Operation, Continuous Flow Pressure Demand Applications
Includes Airline adapter, breathing tube, low pressure connector, waist belt, shoulder strap and full face plug
Model W-3197
Quantity 1 Ea.
Product Type Regulator Kit