Learn More
3M™ Airstream™ Mining Headgear-Mounted Powered Air Purifying Respirator System
Catalog No. 19038880 Shop All 3M Products
Change view
Click to view available options
:
HE filter; Antifog Face Shield; Hard-hat visor
Catalog No.
19-038-880 HE filter; Antifog Face Shield; Hard-hat visor
1 options
Catalog No. 19-038-880 Supplier 3M™ Supplier No. 7100006486
Features positive pressure which help keep out particles, and keep the face shield fog-free.
  • Lightweight and comfortable system. No breathing tubes or hoses are required.
  • 3M Catalog# AS-600LBC
TRUSTED_SUSTAINABILITY
Specifications
Product Type PAPR Hard Hat System
For Use With (Application) For mining, oil and gas industries, assembly, machine operations
Model AS-600LBC