Learn More
MSC 3M AISLE MARKING TAPE
Catalog No. 5000768634
Encompass
This 3M Floor and Aisle Marking Tape is a light-duty, non anti-slip vinyl tape, designed for aisle marking applications. It is 1-1/2 in. wide, 108 ft. long, and 5.2 mil thick.
  • Available in a wide variety of vivid colors for diverse applications.
  • Unique stretch properties enable tape to remain stretched to conform to uneven surfaces without lifting and retracting.
  • One-piece, clean removal from many surfaces which helps reduce clean-up and labor costs.
  • Rubber adhesive provides excellent holding strength on many surfaces.
  • Good abrasion and solvent resistance for long service life.
Catalog No. 50-007-68634 Supplier MSC Supplier No. 06269815
This 3M Floor and Aisle Marking Tape is a light-duty, non anti-slip vinyl tape, designed for aisle marking applications. It is 1-1/2 in. wide, 108 ft. long, and 5.2 mil thick.
  • Available in a wide variety of vivid colors for diverse applications.
  • Unique stretch properties enable tape to remain stretched to conform to uneven surfaces without lifting and retracting.
  • One-piece, clean removal from many surfaces which helps reduce clean-up and labor costs.
  • Rubber adhesive provides excellent holding strength on many surfaces.
  • Good abrasion and solvent resistance for long service life.