Learn More
MSC 3M AISLE MARKING TAPE
Catalog No. 5000803018
Encompass
This vinyl floor and egress tape is designed for aisle marking applications and general traffic areas. It features a solid black color, a 5.2 mil thickness, and is constructed from durable vinyl material with a vinyl adhesive.
  • Light-duty traffic type
  • Non anti-slip surface
  • Black color
  • Vinyl material
  • Vinyl adhesive
  • Aisle marking application
  • Width of 3/4 in
  • Thickness of 5.2 mil
  • Length of 108 ft
  • Solid pattern
Catalog No. 50-008-03018 Supplier MSC Supplier No. 06269708
This vinyl floor and egress tape is designed for aisle marking applications and general traffic areas. It features a solid black color, a 5.2 mil thickness, and is constructed from durable vinyl material with a vinyl adhesive.
  • Light-duty traffic type
  • Non anti-slip surface
  • Black color
  • Vinyl material
  • Vinyl adhesive
  • Aisle marking application
  • Width of 3/4 in
  • Thickness of 5.2 mil
  • Length of 108 ft
  • Solid pattern