Learn More
McKesson General Medical 3M ATTEST BIOLOGICAL INDICATOR
Catalog No. NC0413414
Encompass_Preferred
3Ma, c attesta, c Indicator 1262P Steam 25/b0x This 3Ma, c attesta, c Indicator is for 132 degrees C vacuum assisted or 121 degrees C gravity steam sterilizers. It provides assurance of sterility by challenging the sterilizer with quantifiable, highly resistant spores. Incubation is for 48 hours.
Catalog No. NC0413414 Supplier McKesson General Medical Supplier No. 143644
3Ma, c attesta, c Indicator 1262P Steam 25/b0x This 3Ma, c attesta, c Indicator is for 132 degrees C vacuum assisted or 121 degrees C gravity steam sterilizers. It provides assurance of sterility by challenging the sterilizer with quantifiable, highly resistant spores. Incubation is for 48 hours.