Learn More
Medex Supply 3M Attest Biological Indicator, Rapid Readout, Steam, 3 Hour
SDP
Catalog No. NC3996289
Encompass_Preferred
This dual readout biological indicator system provides rapid and reliable monitoring for steam sterilization processes. It detects Geobacillus stearothermophilus activity to identify sterilization failures, with primary fluorescent results available in 3 hr. and an optional visual color change.
  • Dual readout system for reliable monitoring.
  • Detects Geobacillus stearothermophilus activity.
  • Provides fluorescent results in 3 hr.
  • Optional visual color change from purple to yellow.
  • Compatible with designated auto-readers.
  • Throughput chemical indicator confirms steam exposure (rose to brown/black).
  • Monitors 250°F/121°C gravity and 270°F/132°C vacuum-assisted steam cycles.
  • Meets AAMI test pack recommendations.
  • Color-coded for ease of use and identification.
Catalog No. NC3996289 Supplier Medex Supply Supplier No. 3M12921:200_CS
This dual readout biological indicator system provides rapid and reliable monitoring for steam sterilization processes. It detects Geobacillus stearothermophilus activity to identify sterilization failures, with primary fluorescent results available in 3 hr. and an optional visual color change.
  • Dual readout system for reliable monitoring.
  • Detects Geobacillus stearothermophilus activity.
  • Provides fluorescent results in 3 hr.
  • Optional visual color change from purple to yellow.
  • Compatible with designated auto-readers.
  • Throughput chemical indicator confirms steam exposure (rose to brown/black).
  • Monitors 250°F/121°C gravity and 270°F/132°C vacuum-assisted steam cycles.
  • Meets AAMI test pack recommendations.
  • Color-coded for ease of use and identification.