Learn More
Medplus Services USA Attest Biological Indicators For Steam 270D F/ 132D C Vacuum Assisted or 250D F/ 121D C Gravity Sterilizers, 48 Hour Readout, Brown Cap (100/case)
Supplier:  Medplus Services USA MMM 1262P
Catalog No. NC0344438