Learn More
3M™ Breathe-Easy™ Air Welding System Accessory, Clamp
Catalog No. 19813724 Shop All 3M Products
Change view
Click to view available options
Breathing Tube Clamp
Catalog No.
19-813-724 Breathing Tube Clamp
1 options
This item is not returnable. View return policy
All safety products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product. Misuse may result in sickness or death.
Catalog No. 19-813-724 Supplier 3M™ Supplier No. 7000126381
This item is not returnable. View return policy
All safety products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product. Misuse may result in sickness or death.
Clamp for use with Breathe Easy™ belt-mounted PAPR system breathing tubes.
  • For use with 3M Breathe Easy high-efficiency and supplied air systems
  • 3M Catalog# 529-01-93R01
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
Product Type Clamp
For Use With (Application) Assembly, Cleaning, Electrical, Facility Maintenance, Machine Operation, Painting, Sanding, Welding
For Use With (Equipment) Breathe Easy™ PAPR Systems
Model 5290193R01