Learn More
Enterprise Technology Solutions 3M BX Safety Glass, Silver/Black
SDP
Catalog No. 5000828458
Encompass_Preferred
Sleek, contoured and fully adjustable, BX protective eyewear features a soft adjustable nose bridge, lens angle adjustment, and adjustable-length temples for extended wear. Anti-fog polycarbonate lens provides three lens angles for a cozy fit and absorbs 99.9 percent ultraviolet light. Recommended for assembly, cleaning, electrical, facility maintenance, grinding, machine operations, painting and sanding, these sleek, contoured and adjustable safety glasses help protect your eyes from impact and ultraviolet light. Safety glasses meet the requirements of CSA Z94.3-2007 and the High Impact Requirements of ANSI Z87.1-2003.
Catalog No. 50-008-28458 Supplier Enterprise Technology Solutions Supplier No. MMM113800000020
Sleek, contoured and fully adjustable, BX protective eyewear features a soft adjustable nose bridge, lens angle adjustment, and adjustable-length temples for extended wear. Anti-fog polycarbonate lens provides three lens angles for a cozy fit and absorbs 99.9 percent ultraviolet light. Recommended for assembly, cleaning, electrical, facility maintenance, grinding, machine operations, painting and sanding, these sleek, contoured and adjustable safety glasses help protect your eyes from impact and ultraviolet light. Safety glasses meet the requirements of CSA Z94.3-2007 and the High Impact Requirements of ANSI Z87.1-2003.