Learn More
3M™ PELTOR™ Camelback Gel Sealing Rings
Catalog No. 17000616 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Compatible with Most PELTOR™ Headsets
Catalog No. For Use With (Equipment)
17-000-616 Compatible with Most PELTOR™ Headsets
1 options
Catalog No. 17-000-616 Supplier 3M™ Supplier No. 7000127268
Add to Cart
Add to Cart
Featuring a super-thin polyurethane skin with a double-hump top face design and a silicon gel-filled bladder with a Confor™ foam backing, Camelback Gel sealing rings are compatible with most 3M™ PELTOR™ Headsets.
  • Gel ear cushions for use on a wide variety of 3M™ PELTOR™ Headsets
  • Super-soft foam for enhanced comfort
  • Gel-filled replacement cushions maintain a comfortable, noise-blocking seal around ears
  • Sealing rings maintain original service performance of PELTOR products
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
Cushion Material Polyurethane
Cushion Color Black
For Use With (Equipment) Compatible with Most PELTOR™ Headsets
Model HY80
Product Type Sealing Ring
Material Polyurethane