Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ High Efficiency Cartridge FR-57

Catalog No. 19044570 Shop All 3M Products
Change view
Click to view available options
Quantity:
6/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-044-570 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-044-570 Supplier 3M™ Supplier No. 7000126396
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with 3M™ hood PAPR systems including BE-NM10 PAPR and RBE-L10 PAPR.

  • Filter media uses advanced 3M carbon filtering technology to protect against a wide range of contaminants
  • Filters a variety of chemical warfare agents including certain nerve agents, mustard agents, tear agents, blood agents, phosgene, chloropicrin and diphenylchloroarsine
  • 3M Catalog# 453-03-02R06

Certifications

NIOSH-approved for organic vapor, acid gas (sulfur dioxide, chlorine and hydrogen chloride), ammonia, phosphine, CS, CN and high efficiency filter.

TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type Multi-gas and vapor
Certifications/Compliance NIOSH Approved
Color Magenta/Olive
Product Type FR-57 Respirator Cartridge
For Use With (Application) High efficiency particulate protection in environments containing toxic industrial chemicals
For Use With (Equipment) Breathe Easy™ hooded PAPR systems
Model 453-03-02R06
Quantity 6/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.