Learn More
Enterprise Technology Solutions 3M Cap-Mount Earmuff, f/Hardhat, 24dB NRR, Black
SDP
Catalog No. 5000827911
Encompass_Preferred
Peltor Optime 101 cap-mounted earmuffs feature a stainless steel headband for consistent force and effective hearing protection. They are a nonconductor of direct electric current (dielectric). Noise Reduction Rating (NRR) is 24 decibels. CSA Class A. Cap mounting allows easy use with most hard hats. Design also includes liquid/foam-filled ear cushions and color-coding for compliance sighting. One size fits most. Recommended applications include facility maintenance, grinding, machine operations, sanding, welding, demolition, painting, blasting, assembly and cleaning.
Catalog No. 50-008-27911 Supplier Enterprise Technology Solutions Supplier No. MMMH7P3E
Peltor Optime 101 cap-mounted earmuffs feature a stainless steel headband for consistent force and effective hearing protection. They are a nonconductor of direct electric current (dielectric). Noise Reduction Rating (NRR) is 24 decibels. CSA Class A. Cap mounting allows easy use with most hard hats. Design also includes liquid/foam-filled ear cushions and color-coding for compliance sighting. One size fits most. Recommended applications include facility maintenance, grinding, machine operations, sanding, welding, demolition, painting, blasting, assembly and cleaning.