Learn More
3M™ Cap Mount Hard Hat Headgear
Catalog No. 17987285 Shop All 3M Products
Change view
Click to view available options
Color:
Blue
Catalog No. Color
17-987-285 Blue
1 options
Catalog No. 17-987-285 Supplier 3M™ Supplier No. 7000002291
Add to Cart
Add to Cart
Cap-mounted headgear assembly can be used with any 3M™ Faceshield to help provide face protection. Designed to fit most hard hat accessory slots. High-strength thermoplastic crown provides impact resistance.
3M™ Cap Mount Hard Hat Headgear H18 provides reliable head and face protection products that meet regulated performance and safety requirements. Headgear systems give users a wide choice of faceshield materials, sizes, shapes, and tints for task specific requirements.
  • Versatile headgear assembly can be combined with any 3M faceshield for face protection
  • Mounts to 3M H-700 Hard Hats via the accessory slots
  • Noise Reduction Rating (NRR)* 28 dB. CSA Class AL
  • Requires protective eyewear, sold separately
  • High-strength thermoplastic cap-mount
  • Provides impact resistance and features a unique system that allows window replacement in seconds
  • Meets ANSI Z87.1-2010 requirements
  • 3M Catalog# 82502-00000
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Model 82502-00000
Product Type Headgear Combination
Size Standard (6.5 to 8 in.)
Ventilated Not Ventilated