Learn More
MSC 3M CARTRIDGE
Catalog No. 5000782610
Encompass
The Breathe Easy PAPR Cartridge is a NIOSH-approved filter designed for use with Breathe Easy Powered Air Purifying Respirator (PAPR) Systems. It provides advanced respiratory protection by filtering various contaminants from the air, ensuring a safer breathing environment.
  • For use with Breathe Easy Powered Air Purifying Respirator (PAPR) systems
  • NIOSH-approved
  • Provides multi-gas, organic vapor, and acid gas protection
  • Features a high-efficiency particulate air (HEPA) filter
  • Distinctive magenta/olive color coding for easy identification of protection type
  • Recommended for applications involving welding
  • Case quantity: 6
Catalog No. 50-007-82610 Supplier MSC Supplier No. 04362331
The Breathe Easy PAPR Cartridge is a NIOSH-approved filter designed for use with Breathe Easy Powered Air Purifying Respirator (PAPR) Systems. It provides advanced respiratory protection by filtering various contaminants from the air, ensuring a safer breathing environment.
  • For use with Breathe Easy Powered Air Purifying Respirator (PAPR) systems
  • NIOSH-approved
  • Provides multi-gas, organic vapor, and acid gas protection
  • Features a high-efficiency particulate air (HEPA) filter
  • Distinctive magenta/olive color coding for easy identification of protection type
  • Recommended for applications involving welding
  • Case quantity: 6