Learn More
3M™ Centurion™ Impact Goggles
Catalog No. 19041162 Shop All 3M Products
Change view
Click to view available options
Lens Coating:
Uncoated
Catalog No. Lens Coating
19-041162 Uncoated
1 options
This item is not returnable. View return policy
Catalog No. 19-041162 Supplier 3M™ Supplier No. 7000127564
Add to Cart
Add to Cart
This item is not returnable. View return policy
The durable 3M™ Centurion Impact Safety Goggle is reliable protective eyewear, featuring a wide-angle wraparound frame design with distinct 'side-to-side' peripheral vision and a large lens opening to accommodate prescription and safety eyewear with dust protection.
  • 3M Catalog# 40300-00000-10 , 3M Catalog# 40301-00000-10
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Uncoated
Certifications/Compliance ANSI Z87.1 and CAN / CSA-Z94.3
Lens Tint Clear
Product Type Impact Goggles
For Use With (Equipment) Fits over most glasses
Model 40300-00000-10
Ventilation Details Vertical
Material Polycarbonate
UV Protection UV400