Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Chemical Spill Sorbent Response Pack

Catalog No. 19073207 Shop All 3M Products
Change view
Click to view available options
Capacity (English):
3.75 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-073207 3.75 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-073207 Supplier 3M™ Supplier No. 7000001939
Add to Cart
Add to Cart

Includes

Three mini booms and 10 pads

Designed for use in areas where hazardous chemicals are shipped, stored, or transferred

  • Each 3 in. × 4 ft. (7.6cm × 1.2m) mini boom absorbs up to 1 gal. (3.8L)
  • 10 pads absorb up to 3/4 gal. (2.8L); measure 11 × 13 in. (28 × 33cm) each
  • These safety products must be used in accordance with the user instructions, warnings, and limitations accompanying each product.

  • 3M Catalog# SRPCHEM

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) Sorbing 1 gal./Mini Boom, 3/4 gal./10 Pads
Capacity (Metric) Sorbing 3.8 mL/Mini Boom, 2.8 mL/10 Pads
Capacity (English) 3.75 gal.
Capacity (Metric) 14.2 L
Type Chemical Spill Sorbent Response Pack
For Use With (Application) Chemical spills
Includes (10) 9.25 x 14.05 in pads, (3) 3 in. x 4 ft. booms, 1 contaminated waste bag
Model SRPCHEM
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.