Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Citrus Base Cleaner

Catalog No. 19047248 Shop All 3M Products
Change view
Click to view available options
Quantity:
12 Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-047-248 12 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-047-248 Supplier 3M™ Supplier No. 7000028595
This item is not returnable. View return policy

Solvent-free, noncorrosive, with no petroleum distillates

Heavy-duty, multipurpose aerosol cleaner/degreaser dissolves and flushes away dirt, grease, tar, most noncuring adhesive films, and similar substances. Use on metal parts, motors, tools, printing presses, concrete, or almost any dirty, greasy surface.

  • Solvent-free and noncorrosive
  • Citrus-oil base emulsifies when flushed with water
  • Contains no petroleum distillates
  • Fresh citrus scent
  • 3M Catalog# ITRUSCLNRAERO

Order Info

24 fl. oz. (710mL) can.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Citrus Base Cleaner
Quantity 12 Cs.
Model ITRUSCLNRAERO
For Use With (Application) Flushes away dirt, grease, tar, most noncuring adhesive films, and similar substances
Volume (English) 24 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.