Learn More
3M™ E-A-R™ Classic™ SuperFit™ Ear Plugs
Catalog No. 19042398 Shop All 3M Products
Change view
Click to view available options
Model:
310-1008
311-1126
Corded:
Corded
Uncorded
Size:
Large
Regular
Packaging Type:
Pillow Pk.
Poly bag
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Corded Size Packaging Type
19-042-398 310-1008 Uncorded Large Pillow Pk.
19-042-397 311-1126 Corded Regular Poly bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-042-398 Supplier 3M™ Supplier No. 7000144937
SuperFit™ ear plugs feature a unique ring to help identify how deeply the ear plug is inserted. When minimal to no orange color is visible, the proper insertion is achieved.
  • Orange fitting ring provides a guideline for proper insertion and quick visual compliance check
  • Conforms to the shape of the ear canal
  • Reusable pillow pack or disposable poly bag packaging available
  • Test compatible with 3M™ E-A-Rfit™ Validation System
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Large
For Use With (Application) Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
High Visibility Yes
Model 310-1008
Packaging Type Pillow Pk.
Pairs per Package 2000 Pairs/Cs
Plug Material Foam
Plug Color Yellow, orange
Noise Reduction Rating (NRR) 33 dB
Corded Uncorded
Plug Shape Cylinder
Disposable Single-use
Metal Detectable No