Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Clear Gel Wrist Rest, Beach Design

Catalog No. 19021852 Shop All 3M Products
Change view
Click to view available options
Color:
Beach
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-021-852 Beach
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-021-852 Supplier 3M™ Supplier No. 7010383872
This item is not returnable. View return policy

Unique combination of soothing gel and great-feeling fabric increases comfort and can improve the performance of your optical mouse

  • Helps maintain comfort throughout the day by helping keep wrist in a neutral position
  • Helps reduce pressure points
  • Patented gel technology
  • Non-slip to stay in place
  • Clear gel with beach design
  • 3M Catalog# WR308BH

TRUSTED_SUSTAINABILITY

Specifications

Color Beach
Product Type Gel Wrist Rest
Length (Metric) 45.7 cm
Length (English) 18 in.
Size Compact
For Use With (Application) Inhibits the growth of microorganisms such as bacteria and encourages neutral wrist posture
Includes Non-skid base, clear gel
Material Foam
Model WR308BH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.