Learn More
3M™ Communication Headsets: Microphones
Catalog No. 191202641 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Com-Tac or Sweet-Tac Headsets
Catalog No. For Use With (Equipment)
19-120-2641 Com-Tac or Sweet-Tac Headsets
1 options
Catalog No. 19-120-2641 Supplier 3M™ Supplier No. 7000127275
This throat microphone allows for crisp, reliable communication in the two-way communication headsets of the 3M™ PELTOR™ Series, specifically designed for industrial users in high-noise environments.
  • Throat microphone for use on the 3M™ PELTOR™ COMTAC™ III Communication Headsets
  • Can be used as an alternative to the boom microphone
  • A practical solution when boom microphones are ineffective or can’t be used, such as when wearing other personal protective equipment or headborne objects
  • Picks up vibrations from user’s voice box for voice transmission
  • Effective in noisy environments
  • Offers stretchable strap with adjustments for different neck sizes
  • Plugs into the same auxiliary port as the flexible boom mic
  • 3M Catalog# MT90
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Com-Tac or Sweet-Tac Headsets
Model MT90
Product Type Throat microphon